N-Succinimidyl Methacrylate ≥98.0%

Supplier: TCI America

S0812-25G S0812-5G
TCS0812-25GEA 996.98 CAD
TCS0812-25G TCS0812-5G
N-Succinimidyl Methacrylate ≥98.0%
N-Succinimidyl Methacrylate

CAS Number: 38862-25-8
Molecular Formula: C8H9NO4
Molecular Weight: 183.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 104
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide

Formula: C₈H₉NO₄
Melting Pt: 104 °C
MDL Number: MFCD15072193
CAS Number: 38862-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c069c1cc8d9979b60afb55665114e550269a19d&vid=5c069c1cc8d9979b60afb55665114e550269a19d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR