Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate hydrate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonic acid Sodium Salt, 3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonic acid Sodium Salt, 3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt, TOOS, 3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulphonic acid sodium salt
CAS Number: 82692-93-1
MDL Number: MFCD00040641
Molecular Formula: C12H19NO4S
Molecular Weight: 295.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonic Acid Sodium Salt, TOOS
Formula:
C₁₂H₁₉NO₄S MW: 295.33 g/mol |
MDL Number:
MFCD00040641 CAS Number: 82692-93-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...