9,9'-Spirobi[9H-fluorene] ≥98.0%

Supplier: TCI America

Synonyms: 9,9'-Spirobi[fluorene], 9,9'-Spirobifluorene

S0800-1G S0800-5G
TCS0800-1GEA 93.22 CAD
TCS0800-1G TCS0800-5G
9,9'-Spirobi[9H-fluorene] ≥98.0%
9,9'-Spirobi[9H-fluorene]

CAS Number: 159-66-0
MDL Number: MFCD08704216
Molecular Formula: C25H16
Molecular Weight: 316.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 202

Formula: C₂₅H₁₆
Melting Pt: 202 °C
MDL Number: MFCD08704216
CAS Number: 159-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e341404f6afc48283bdff5d0406bb2931ce972a&vid=9e341404f6afc48283bdff5d0406bb2931ce972a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR