3-Amino-4-hydroxyphenyl methyl sulfone ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A2198-25G A2198-5G
TCA2198-25GEA 113.76 CAD
TCA2198-25G TCA2198-5G
3-Amino-4-hydroxyphenyl methyl sulfone ≥97.0% (by HPLC, titration analysis)
3-Amino-4-hydroxyphenyl methyl sulfone

CAS Number: 98-30-6
MDL Number: MFCD00719450
Molecular Formula: C7H9NO3S
Molecular Weight: 187.21
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158


Other Product Names / Synonyms: 2-Amino-4-(methylsulfonyl)phenol, 2-Hydroxy-5-(methylsulfonyl)aniline

Formula: C₇H₉NO₃S
Melting Pt: 158 °C
MDL Number: MFCD00719450
CAS Number: 98-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42aaa07a3b425fa5ba790bd0e6080193ca82bdd8&vid=42aaa07a3b425fa5ba790bd0e6080193ca82bdd8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR