S-Sodium ethanethiosulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0533-500G S0533-25G
TCS0533-500GEA 1535.76 CAD
TCS0533-500G TCS0533-25G
S-Sodium ethanethiosulfonate ≥98.0% (by HPLC, titration analysis)
S-Sodium ethanethiosulfonate

CAS Number: 31999-88-9
Molecular Formula: C2H6O2S2
Molecular Weight: 148.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Ethanethiosulfonic Acid S-Sodium Salt

Formula: C₂H₆O₂S₂
CAS Number: 31999-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2085b5f6626aebe6fb7518e7a3129857f75e9053&vid=2085b5f6626aebe6fb7518e7a3129857f75e9053&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR