Solanesol ≥93.0%

Supplier: TCI America

Synonyms: (2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-Nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaen-1-ol

S0525-25G S0525-5G
TCS0525-25GEA 644.64 CAD
TCS0525-25G TCS0525-5G
Solanesol ≥93.0%
Solanesol

CAS Number: 13190-97-1
MDL Number: MFCD00070279
Molecular Formula: C45H74O
Molecular Weight: 631.09
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 42
Storage Temperature: 0-10°C

Formula: C₄₅H₇₄O
Melting Pt: 42 °C
MDL Number: MFCD00070279
CAS Number: 13190-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8da8fd2106c45515f5b92e2faeeaa36788e55316&vid=8da8fd2106c45515f5b92e2faeeaa36788e55316&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR