Sodium-4-(2-bromoethyl)benzenesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0484-25G S0484-5G
TCS0484-25GEA 500.86 CAD
TCS0484-25G TCS0484-5G
Sodium-4-(2-bromoethyl)benzenesulfonate ≥98.0% (by titrimetric analysis)
Sodium-4-(2-bromoethyl)benzenesulfonate

CAS Number: 65036-65-9
MDL Number: MFCD06797167
Molecular Formula: C8H9BrO3S
Molecular Weight: 287.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(2-Bromoethyl)benzenesulfonic Acid Sodium Salt

Formula: C₈H₉BrO₃S
MDL Number: MFCD06797167
CAS Number: 65036-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b33aa4f9f54fb3d2670752bbc5054e204d8cf893&vid=b33aa4f9f54fb3d2670752bbc5054e204d8cf893&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR