Methyl succinamate ≥98.0%

Supplier: TCI America

S0479-25G S0479-5G
TCS0479-25GEA 226.41 CAD
TCS0479-25G TCS0479-5G
Methyl succinamate ≥98.0%
Methyl succinamate

CAS Number: 53171-39-4
Molecular Formula: C5H9NO3
Molecular Weight: 131.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 118
Melting point (°C): 89


Other Product Names / Synonyms: Succinamic Acid Methyl Ester

Formula: C₅H₉NO₃
Boiling Pt: 118 °C
Melting Pt: 89 °C
MDL Number: MFCD08276419
CAS Number: 53171-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fa33b8e09734244e3c9d6889e61ef0c2e4e5000&vid=0fa33b8e09734244e3c9d6889e61ef0c2e4e5000&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR