O-Acetyl-N-carbobenzoxyhydroxylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Acetoxycarbamic Acid Benzyl Ester, Benzyl N-Acetoxycarbamate, O-Acetyl-N-Cbz-hydroxylamine

A2193-1G A2193-5G
TCA2193-1GEA 150.1 CAD
TCA2193-1G TCA2193-5G
O-Acetyl-N-carbobenzoxyhydroxylamine ≥98.0% (by titrimetric analysis)
O-Acetyl-N-carbobenzoxyhydroxylamine

CAS Number: 180798-01-0
Molecular Formula: C10H11NO4
Molecular Weight: 209.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: O-Acetyl-N-Cbz-hydroxylamine, N-Acetoxycarbamic Acid Benzyl Ester, Benzyl N-Acetoxycarbamate

Formula: C₁₀H₁₁NO₄
MW: 209.20 g/mol
CAS Number: 180798-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa399b6259fba96405f7b5428b631bd83bf3c15&vid=6fa399b6259fba96405f7b5428b631bd83bf3c15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR