4-Sulfothiacalix[4]arene sodium salt ≥98.0% (by HPLC)

Supplier: TCI America

S0477-5G S0477-1G
TCS0477-5GEA 597.24 CAD
TCS0477-5G TCS0477-1G
4-Sulfothiacalix[4]arene sodium salt ≥98.0% (by HPLC)
4-Sulfothiacalix[4]arene sodium salt

CAS Number: 211561-04-5
MDL Number: MFCD06797166
Molecular Formula: C24H16O16S8
Molecular Weight: 904.78
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 390


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetrasodium Tetrathiacalix[4]arenetetrasulfonate, Thiacalix[4]arenetetrasulfonic Acid Tetrasodium Salt

Formula: C₂₄H₁₆O₁₆S₈
Melting Pt: 390 °C
MDL Number: MFCD06797166
CAS Number: 211561-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8234558644012d6519a57d45e518f226444e4a1b&vid=8234558644012d6519a57d45e518f226444e4a1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR