4-Sulfocalix[8]arene hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0471-5G S0471-1G
TCS0471-5GEA 1256.1 CAD
TCS0471-5G TCS0471-1G
4-Sulfocalix[8]arene hydrate ≥98.0% (by HPLC, titration analysis)
4-Sulfocalix[8]arene hydrate

CAS Number: 137407-62-6
MDL Number: MFCD00917032
Molecular Formula: C56H48O32S8
Molecular Weight: 1489.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White

Formula: C₅₆H₄₈O₃₂S₈
MDL Number: MFCD00917032
CAS Number: 137407-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c9976e369b2b9e342e80d2419bc658d847e9a21&vid=7c9976e369b2b9e342e80d2419bc658d847e9a21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR