4-Sulfocalix[4]arene hydrate ≥94.0% (by titrimetric analysis)
Supplier: TCI America
S0469-5G
S0469-1G
TCS0469-5GEA
1414.37
CAD
TCS0469-5G
TCS0469-1G
4-Sulfocalix[4]arene hydrate ≥94.0% (by titrimetric analysis)
4-Sulfocalix[4]arene hydrate
CAS Number: 112269-92-8
MDL Number: MFCD03095486
Molecular Formula: C28H24O16S4
Molecular Weight: 744.72
Purity/Analysis Method: >94.0% (T)
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetrahydroxycalix[4]arenetetrasulfonic Acid, Tetrasulfo(tetrahydroxy)calix[4]arene
Formula:
C₂₈H₂₄O₁₆S₄ |
MDL Number:
MFCD03095486 CAS Number: 112269-92-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...