2-Aminoethyl hydrogen sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0445-500G S0445-25G
TCS0445-500GEA 199.08 CAD
TCS0445-500G TCS0445-025G
2-Aminoethyl hydrogen sulfate ≥98.0% (by titrimetric analysis)
2-Aminoethyl hydrogen sulphate

CAS Number: 926-39-6
MDL Number: MFCD00008179
Molecular Formula: C2H7NO4S
Molecular Weight: 141.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 279


Other Product Names / Synonyms: Sulfuric Acid Mono(2-aminoethyl) Ester

Formula: C₂H₇NO₄S
Melting Pt: 279 °C
MDL Number: MFCD00008179
CAS Number: 926-39-6
EINECS: 213-135-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4021934fd115d60ae2e6ef8751580de9f11f2aa&vid=d4021934fd115d60ae2e6ef8751580de9f11f2aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR