SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0%

Supplier: TCI America

Synonyms: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

S0431-5G S0431-1G
TCS0431-5GEA 2842.42 CAD
TCS0431-5G TCS0431-001G
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0%
SATA (N-Succinimidyl S-Acetylthioglycolate)

[Cross-linking Reagent]
CAS Number: 76931-93-6
MDL Number: MFCD00036891
Molecular Formula: C8H9NO5S
Molecular Weight: 231.22
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 97
Storage Temperature: <0°C


Product Categories: Bioscience, Functionality Creating Reagents
Other Product Names / Synonyms: S-Acetylthioglycolic Acid N-Succinimidyl Ester

Formula: C₈H₉NO₅S
MW: 231.22 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00036891
CAS Number: 76931-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a29b789ba65419b2f4ced3cfb80f85c690391d0&vid=4a29b789ba65419b2f4ced3cfb80f85c690391d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR