N-Succinimidyl 4-maleimidobutyrate (GMBS) ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
S0399-1G
S0399-100MG
TCS0399-001GEA
311.26
CAD
TCS0399-001G
TCS0399-100MG
N-Succinimidyl 4-maleimidobutyrate (GMBS) ≥98.0% (by HPLC, total nitrogen)
GMBS (N-Succinimidyl 4-maleimidobutyrate)
[Cross-linking Reagent]
CAS Number: 80307-12-6
MDL Number: MFCD00036817
Molecular Formula: C12H12N2O6
Molecular Weight: 280.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 130
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 4-Maleimidobutyric Acid N-Succinimidyl Ester, N-(4-Maleimidobutyryloxy)succinimide, GMBS
Formula:
C₁₂H₁₂N₂O₆ Melting Pt: 130 °C |
MDL Number:
MFCD00036817 CAS Number: 80307-12-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...