ɑ-Amino-γ-butyrolactone hydrobromide 98,0
Supplier: TCI America
Synonyms:
DL-Homoserine lactone hydrobromide
A0283-1G
A0283-5G
TCA0283-001GEA
50.56
CAD
TCA0283-001G
TCA0283-005G
ɑ-Amino-γ-butyrolactone hydrobromide 98,0
alpha-Amino-gamma-butyrolactone hydrobromide
CAS Number: 6305-38-0
MDL Number: MFCD00012723
Molecular Formula: C4H7NO2
Molecular Weight: 182.02
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 237
Formula:
C₄H₇NO₂ MW: 182.02 g/mol Melting Pt: 237 °C |
MDL Number:
MFCD00012723 CAS Number: 6305-38-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...