N-Succinimidyl 3-Maleimidobenzoate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

S0398-1G S0398-100MG
TCS0398-001GEA 330.54 CAD
TCS0398-001G TCS0398-100MG
N-Succinimidyl 3-Maleimidobenzoate ≥98.0% (by HPLC, total nitrogen)
MBS (N-Succinimidyl 3-maleimidobenzoate)

[Cross-linking Reagent]
CAS Number: 58626-38-3
MDL Number: MFCD00005514
Molecular Formula: C15H10N2O6
Molecular Weight: 314.25
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 181
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 3-Maleimidobenzoic Acid N-Succinimidyl Ester, N-(3-Maleimidobenzoyloxy)succinimide, MBS

Formula: C₁₅H₁₀N₂O₆
Melting Pt: 181 °C
MDL Number: MFCD00005514
CAS Number: 58626-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d9b284f617302638c29393eb3993b9f9dad05a7&vid=9d9b284f617302638c29393eb3993b9f9dad05a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR