Monomethyl sebacate ≥98.0%

Supplier: TCI America

Synonyms: 10-Methoxy-10-oxodecanoic acid, Methyl Hydrogen Sebacate, Decanedioic Acid Monomethyl Ester, Sebacic Acid Monomethyl Ester

S0395-5G S0395-25G
TCS0395-005GEA 26.86 CAD
TCS0395-005G TCS0395-25G
Monomethyl sebacate ≥98.0%
Monomethyl sebacate

CAS Number: 818-88-2
MDL Number: MFCD00004439
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 42
Flash Point (°C): 110


Other Product Names / Synonyms: Decanedioic Acid Monomethyl Ester, Methyl Hydrogen Sebacate, Sebacic Acid Monomethyl Ester

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Melting Pt: 42 °C
Flash Pt: 110 °C
MDL Number: MFCD00004439
CAS Number: 818-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc2747c4a8aa8cd9aa893a8487a512aab4e6280f&vid=bc2747c4a8aa8cd9aa893a8487a512aab4e6280f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR