1-(3-sulfopropyl)pyridinium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

S0391-25G S0391-250G
TCS0391-025GEA 30.56 CAD
TCS0391-025G TCS0391-250G
1-(3-sulfopropyl)pyridinium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen)
1-(3-sulfopropyl)pyridinium hydroxide inner salt

CAS Number: 15471-17-7
MDL Number: MFCD00064468
Molecular Formula: C8H11NO3S
Molecular Weight: 201.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 279
Flash Point (°C): 160


Other Product Names / Synonyms: 3-(1-Pyridinio)propanesulfonate

Formula: C₈H₁₁NO₃S
Melting Pt: 279 °C
Flash Pt: 160 °C
MDL Number: MFCD00064468
CAS Number: 15471-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d729438e35259b127c700e5ec61c7f2d1544b66&vid=6d729438e35259b127c700e5ec61c7f2d1544b66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR