Succinaldehyde bis(dimethyl acetal) ≥98.0%

Supplier: TCI America

Synonyms: Succinaldehyde Tetramethyl Acetal, 1,1,4,4-Tetramethoxybutane

S0371-25G S0371-5G
TCS0371-025GEA 440.82 CAD
TCS0371-025G TCS0371-005G
Succinaldehyde bis(dimethyl acetal) ≥98.0%
Succinaldehyde bis(dimethyl acetal)

CAS Number: 6922-39-0
MDL Number: MFCD00127971
Molecular Formula: C8H18O4
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 89
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Succinaldehyde Tetramethyl Acetal, 1,1,4,4-Tetramethoxybutane

Formula: C₈H₁₈O₄
Boiling Pt: 89 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00127971
CAS Number: 6922-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d50b680743ed2bd73db3eb7dc35b5e594c47a9a2&vid=d50b680743ed2bd73db3eb7dc35b5e594c47a9a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR