Scopoletin (7-hydroxy-6-methoxycumarin) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

S0367-1G S0367-100MG
TCS0367-001GEA 1027 CAD
TCS0367-001G TCS0367-100MG
Scopoletin (7-hydroxy-6-methoxycumarin) ≥98.0% (by HPLC, titration analysis)
Scopoletin (7-Hydroxy-6-methoxycumarin)

CAS Number: 92-61-5
MDL Number: MFCD00006872
Molecular Formula: C10H8O4
Molecular Weight: 192.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 207


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Esculetin-6-methyl Ether, 7-Hydroxy-6-methoxycoumarin

Formula: C₁₀H₈O₄
MW: 192.1681 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00006872
CAS Number: 92-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95fa88669f6f4b5e23f3bea5cd689cab57b1a937&vid=95fa88669f6f4b5e23f3bea5cd689cab57b1a937&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR