Glycidyl stearate ≥96.0% (by titrimetric analysis)

Supplier: TCI America

S0366-5G S0366-25G
TCS0366-005GEA 264.52 CAD
TCS0366-005G TCS0366-25G
Glycidyl stearate ≥96.0% (by titrimetric analysis)
Glycidyl stearate

CAS Number: 7460-84-6
MDL Number: MFCD00059110
Molecular Formula: C21H40O3
Molecular Weight: 340.55
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 193
Melting point (°C): 53


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Stearic Acid Glycidyl Ester

Formula: C₂₁H₄₀O₃
Boiling Pt: 193 °C
Melting Pt: 53 °C
MDL Number: MFCD00059110
CAS Number: 7460-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a4a2c44a1dd040fc85a5de5b70d0a32539cf6dd&vid=7a4a2c44a1dd040fc85a5de5b70d0a32539cf6dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR