4-tert-Butylphenyl salicylate ≥98.0%
Supplier: TCI America
S0274-25G
S0274-500G
TCS0274-025GEA
50.56
CAD
TCS0274-025G
TCS0274-500G
4-tert-Butylphenyl salicylate ≥98.0%
4-tert-Butylphenyl salicylate
CAS Number: 87-18-3
MDL Number: MFCD00045759
Molecular Formula: C17H18O3
Molecular Weight: 270.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 63
Other Product Names / Synonyms: Salicylic Acid 4-tert-Butylphenyl Ester
Formula:
C₁₇H₁₈O₃ Melting Pt: 63 °C |
MDL Number:
MFCD00045759 CAS Number: 87-18-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...