Disodium sebacate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium decanedioate, Decanedioic Acid Disodium Salt, Disodium Decanedioate, Disodium Sebacate, Sebacic Acid Disodium Salt

S0270-25G S0270-500G
TCS0270-025GEA 115.34 CAD
TCS0270-025G TCS0270-500G
Disodium sebacate ≥97.0% (by titrimetric analysis)
Disodium sebacate

CAS Number: 17265-14-4
MDL Number: MFCD00070501
Molecular Formula: C10H18O4
Molecular Weight: 246.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Decanedioic Acid Disodium Salt, Sebacic Acid Disodium Salt, Disodium Decanedioate

Formula: C₁₀H₁₈O₄
MW: 246.21 g/mol
MDL Number: MFCD00070501
CAS Number: 17265-14-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f789f9297d4ac7073531c76c01294c4521ab3247&vid=f789f9297d4ac7073531c76c01294c4521ab3247&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR