Diisopropyl succinate ≥99.0%
Supplier: TCI America
S0269-25ML
S0269-500ML
TCS0269-025MLEA
74.26
CAD
TCS0269-025ML
TCS0269-500ML
Diisopropyl succinate ≥99.0%
Diisopropyl succinate
CAS Number: 924-88-9
MDL Number: MFCD00059360
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 228
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Succinic Acid Diisopropyl Ester
Formula:
C₁₀H₁₈O₄ Boiling Pt: 228 °C Density: 0.99 (20 °C) |
MDL Number:
MFCD00059360 CAS Number: 924-88-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...