Iron(III) stearate
Supplier: TCI America
S0256-25G
S0256-100G
TCS0256-025GEA
88.39
CAD
TCS0256-025G
TCS0256-100G
Iron(III) stearate
Iron(III) stearate
CAS Number: 555-36-2
MDL Number: MFCD00050716
Molecular Formula: C18H36O2
Molecular Weight: 906.27
Form: Crystal
Color: Very Pale Yellow
Flash Point (°C): 150
Other Product Names / Synonyms: Ferric Stearate, Stearic Acid Iron(III) Salt
Formula:
C₁₈H₃₆O₂ Flash Pt: 150 °C |
MDL Number:
MFCD00050716 CAS Number: 555-36-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...