4-(Trifluoromethyl)anthranilic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A2175-25G A2175-5G
TCA2175-25GEA 529.3 CAD
TCA2175-25G TCA2175-5G
4-(Trifluoromethyl)anthranilic acid ≥97.0% (by HPLC, titration analysis)
4-(Trifluoromethyl)anthranilic acid

CAS Number: 402-13-1
MDL Number: MFCD00519334
Molecular Formula: C8H6F3NO2
Molecular Weight: 205.14
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)anthranilic Acid

Formula: C₈H₆F₃NO₂
Melting Pt: 175 °C
MDL Number: MFCD00519334
CAS Number: 402-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=234c407e4f1c5ed40f541e715fd9009764e9b98b&vid=234c407e4f1c5ed40f541e715fd9009764e9b98b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR