Succinaldehyde disodium bisulfite ≥92.0% (by titrimetric analysis)

Supplier: TCI America

S0216-500G S0216-25G
TCS0216-500GEA 474 CAD
TCS0216-500G TCS0216-025G
Succinaldehyde disodium bisulfite ≥92.0% (by titrimetric analysis)
Succinaldehyde disodium bisulfite

CAS Number: 5450-96-4
MDL Number: MFCD00024982
Molecular Formula: C4H10O8S2
Molecular Weight: 294.20
Purity/Analysis Method: >92.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Disodium 1,4-Dihydroxy-1,4-butanedisulfonate

Formula: C₄H₁₀O₈S₂
MDL Number: MFCD00024982
CAS Number: 5450-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdfd8f9cb867f1074ae25b97c44c3551e9a35961&vid=bdfd8f9cb867f1074ae25b97c44c3551e9a35961&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR