Dibenzyl sebacate ≥98.0%

Supplier: TCI America

Synonyms: Decanedioic Acid Dibenzyl Ester, Sebacic Acid Dibenzyl Ester

S0147-25G S0147-5G
TCS0147-025GEA 317.42 CAD
TCS0147-025G TCS0147-5G
Dibenzyl sebacate ≥98.0%
Dibenzyl sebacate

CAS Number: 140-24-9
MDL Number: MFCD00039640
Molecular Formula: C24H30O4
Molecular Weight: 382.50
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 232
Freezing point (°C): 27


Other Product Names / Synonyms: Decanedioic Acid Dibenzyl Ester, Sebacic Acid Dibenzyl Ester

Formula: C₂₄H₃₀O₄
MW: 382.50 g/mol
Flash Pt: 232 °C
MDL Number: MFCD00039640
CAS Number: 140-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52a8db11c09e0f1048efe2cf83abe451a74300d&vid=d52a8db11c09e0f1048efe2cf83abe451a74300d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR