2-Sulfobenzoic cyclic anhydride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Sulfobenzoic anhydride, 2-sulphobenzoic anhydride, 2-sulphobenzoic acid cyclic anhydride, 2-Sulfobenzoic acid cyclic anhydride
S0124-25G
S0124-500G
S0124-100G
TCS0124-025GEA
205.78
CAD
TCS0124-025G
TCS0124-500G
TCS0124-100G
2-Sulfobenzoic cyclic anhydride ≥97.0% (by titrimetric analysis)
2-Sulphobenzoic cyclic anhydride
CAS Number: 81-08-3
MDL Number: MFCD00005879
Molecular Formula: C7H4O4S
Molecular Weight: 184.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 125
Formula:
C₇H₄O₄S MW: 184.17 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00005879 CAS Number: 81-08-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...