Monosodium succinate ≥98.0%

Supplier: TCI America

Synonyms: Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt, Sodium Bisuccinate

S0106-25G S0106-500G
TCS0106-025GEA 67.26 CAD
TCS0106-025G TCS0106-500G
Monosodium succinate ≥98.0%
Monosodium succinate

CAS Number: 2922-54-5
MDL Number: MFCD00050993
Molecular Formula: C4H6O4
Molecular Weight: 140.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Sodium Bisuccinate, Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt

Formula: C₄H₆O₄
MW: 140.07 g/mol
MDL Number: MFCD00050993
CAS Number: 2922-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54a0c51f5af7f9993292eeb6091b76b303ddc88e&vid=54a0c51f5af7f9993292eeb6091b76b303ddc88e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR