Nɑ-Acetyl-L-lysine 97,0
Supplier: TCI America
Synonyms:
Nalpha-Acetyl-L-lysine, Ac-Lys-OH
A2171-1G
A2171-5G
TCA2171-1GEA
102.54
CAD
TCA2171-1G
TCA2171-5G
Nɑ-Acetyl-L-lysine 97,0
Nalpha-Acetyl-L-lysine
CAS Number: 1946-82-3
MDL Number: MFCD00008233
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 254
Specific rotation [a]20/D: 4.0 deg (C=4, H2O)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Lys-OH
Formula:
C₈H₁₆N₂O₃ MW: 188.23 g/mol Melting Pt: 254 °C |
MDL Number:
MFCD00008233 CAS Number: 1946-82-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...