Stearamide ≥90.0%
Supplier: TCI America
Synonyms:
Octadecanamide, Stearic acid amide
S0075-25G
S0075-250G
TCS0075-025GEA
58.22
CAD
TCS0075-025G
TCS0075-250G
Stearamide ≥90.0%
Stearamide
CAS Number: 124-26-5
MDL Number: MFCD00008038
Molecular Formula: C18H37NO
Molecular Weight: 283.50
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 109
Flash Point (°C): 197
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Octadecanamide
Formula:
C₁₈H₃₇NO Melting Pt: 109 °C Flash Pt: 197 °C |
MDL Number:
MFCD00008038 CAS Number: 124-26-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...