Spermidine phosphate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-(3-Aminopropyl)-1,4-butanediamine Phosphate, N-(3-Aminopropyl)tetramethylenediamine Phosphate
S0068-1G
S0068-5G
TCS0068-001GEA
97.49
CAD
TCS0068-001G
TCS0068-005G
Spermidine phosphate ≥98.0% (by titrimetric analysis)
Spermidine phosphate
CAS Number: 49721-50-8
MDL Number: MFCD00043283
Molecular Formula: C7H19N3
Molecular Weight: 439.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Bioscience, Polyamines
Other Product Names / Synonyms: N-(3-Aminopropyl)-1,4-butanediamine Phosphate, N-(3-Aminopropyl)tetramethylenediamine Phosphate
Formula:
C₇H₁₉N₃ MW: 439.23 g/mol |
MDL Number:
MFCD00043283 CAS Number: 49721-50-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...