Span® 85

Supplier: TCI America

S0064-25G S0064-500G
TCS0064-025GEA 44.78 CAD
TCS0064-025G TCS0064-500G
Span® 85
Span® 85

CAS Number: 26266-58-0
MDL Number: MFCD00133820
Form: Clear Liquid
Color: Yellow
Melting point (°C): -23
Flash Point (°C): 253
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Arlacel 85, Sorbitan Trioleate

Formula: C₆₀H₁₀₈O₈
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: –23 °C
Density: 0.95 (20 °C)
Flash Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00133820
CAS Number: 26266-58-0
EINECS: 247-569-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR