Span® 80
Supplier: TCI America
S0060-25G
S0060-500G
TCS0060-025GEA
56.31
CAD
TCS0060-025G
TCS0060-500G
Span® 80
Span® 80
CAS Number: 1338-43-8
MDL Number: MFCD00080948
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 113
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Arlacel 80, Sorbitan Monooleate
Formula:
C₂₄H₄₄O₆ Density: 0.99 (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080948 CAS Number: 1338-43-8 EINECS: 215-665-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...