D-(+)-Sucrose octaacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0052-25G S0052-500G
TCS0052-025GEA 27.33 CAD
TCS0052-025G TCS0052-500G
D-(+)-Sucrose octaacetate ≥98.0% (by titrimetric analysis)
D(+)-Sucrose octaacetate

CAS Number: 126-14-7
MDL Number: MFCD00006623
Molecular Formula: C28H38O19
Molecular Weight: 678.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 86
Specific rotation [a]20/D: 60 deg (C=2, EtOH)


Product Categories: Bioscience, Disaccharides, O-Substituted Sugars
Other Product Names / Synonyms: D-(+)-Saccharose Octaacetate, Octa-O-acetyl D-(+)-Saccharose, D-(+)-Sucrose Octaacetate

Formula: C₂₈H₃₈O₁₉
Melting Pt: 86 °C
Density: 1.257 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006623
CAS Number: 126-14-7
EINECS: 204-772-1
Merck Index: 12,09052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef8bcfc89738503f5e1a3e59082a2c8af41f513&vid=5ef8bcfc89738503f5e1a3e59082a2c8af41f513&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR