DL-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0281-1G A0281-25G A0281-5G
TCA0281-001GEA 39.5 CAD
TCA0281-001G TCA0281-025G TCA0281-5G
DL-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
DL-3-Aminobutyric acid

CAS Number: 541-48-0
MDL Number: MFCD00008087
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 194

Formula: C₄H₉NO₂
Melting Pt: 194 °C
MDL Number: MFCD00008087
CAS Number: 541-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0555aae9bc9c217195be49d61722032ae1aee927&vid=0555aae9bc9c217195be49d61722032ae1aee927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR