Butyl ricinoleate ≥74.0%

Supplier: TCI America

R0045-25ML R0045-500ML
TCR0045-025MLEA 26.86 CAD
TCR0045-025ML TCR0045-500ML
Butyl ricinoleate ≥74.0%
Butyl ricinoleate

CAS Number: 151-13-3
MDL Number: MFCD00136031
Molecular Formula: C22H42O3
Molecular Weight: 354.58
Purity/Analysis Method: >74.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 275
Flash Point (°C): 115
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester

Formula: C₂₂H₄₂O₃
Boiling Pt: 275 °C
Density: 0.91 (20 °C)
Flash Pt: 115 °C
MDL Number: MFCD00136031
CAS Number: 151-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f59f407ee1287ddd351bd1e9fef649cf1fdd412&vid=2f59f407ee1287ddd351bd1e9fef649cf1fdd412&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 74.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR