Sodium ricinolate ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ricinolic acid Sodium Salt, Sodium (R,Z)-12-hydroxyoctadec-9-enoate, Ricinoleic acid Sodium Salt

R0030-500G R0030-25G
TCR0030-500GEA 532.3 CAD
TCR0030-500G TCR0030-025G
Sodium ricinolate ≥93.0% (by titrimetric analysis)
Sodium ricinolate

CAS Number: 5323-95-5
MDL Number: MFCD00050760
Molecular Formula: C18H34O3
Molecular Weight: 320.45
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ricinolic Acid Sodium Salt, Ricinoleic Acid Sodium Salt

Formula: C₁₈H₃₄O₃
MW: 320.45 g/mol
MDL Number: MFCD00050760
CAS Number: 5323-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2695fea90bd90754991716fd90bb245aeae86a3a&vid=2695fea90bd90754991716fd90bb245aeae86a3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR