Methyl ricinoleate ≥75.0%

Supplier: TCI America

R0029-25ML R0029-500ML
TCR0029-025MLEA 69.11 CAD
TCR0029-025ML TCR0029-500ML
Methyl ricinoleate ≥75.0%
Methyl ricinoleate

CAS Number: 141-24-2
MDL Number: MFCD00046712
Molecular Formula: C19H36O3
Molecular Weight: 312.49
Purity/Analysis Method: >75.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 209
Specific Gravity (20/20): 0.92


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ricinoleic Acid Methyl Ester, Ricinolic Acid Methyl Ester

Formula: C₁₉H₃₆O₃
Density: 0.92 (20 °C)
Flash Pt: 209 °C
MDL Number: MFCD00046712
CAS Number: 141-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60cb94a77c16fa9d6c1514492c50eee0c884b521&vid=60cb94a77c16fa9d6c1514492c50eee0c884b521&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR