Ricinoleic acid ≥80.0%
Supplier: TCI America
Synonyms:
Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid
R0027-25ML
R0027-500ML
TCR0027-025MLEA
57.02
CAD
TCR0027-025ML
TCR0027-500ML
Ricinoleic acid ≥80.0%
Ricinoleic acid
CAS Number: 141-22-0
MDL Number: MFCD00084840
Molecular Formula: C18H34O3
Molecular Weight: 298.47
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 248
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: Ricinolic Acid
Formula:
C₁₈H₃₄O₃ MW: 298.47 g/mol Density: 0.94 (20 °C) Flash Pt: 248 °C |
MDL Number:
MFCD00084840 CAS Number: 141-22-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...