N-Acetyl-D-mannosamine monohydrate ≥98.0%

Supplier: TCI America

A2160-1G A2160-5G
TCA2160-1GEA 311.75 CAD
TCA2160-1G TCA2160-5G
N-Acetyl-D-mannosamine monohydrate ≥98.0%
N-Acetyl-D-mannosamine monohydrate

CAS Number: 3615-17-6
MDL Number: MFCD00136044
Molecular Formula: C8H15NO6
Molecular Weight: 221.21
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 125


Product Categories: Bioscience, Mannosamine
Other Product Names / Synonyms: 2-Acetamido-2-deoxy-D-mannopyranose

Formula: C₈H₁₅NO₆·H₂O
MW: 239.24 g/mol (221.22 anhy)
Melting Pt: 125 °C
MDL Number: MFCD00149493
CAS Number: 1071625-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=125d113c0f415991aad24e5e250175d49a077bd1&vid=125d113c0f415991aad24e5e250175d49a077bd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR