Resorufin ≥95.0% (by HPLC)

Supplier: TCI America

R0012-5G R0012-1G
TCR0012-005GEA 1183.42 CAD
TCR0012-005G TCR0012-1G
Resorufin ≥95.0% (by HPLC)
Resorufin

CAS Number: 635-78-9
MDL Number: MFCD00128991
Molecular Formula: C12H7NO3
Molecular Weight: 213.19
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Deep Green

Formula: C₁₂H₇NO₃
MDL Number: MFCD00128991
CAS Number: 635-78-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=136f3009600b6c3abccdf5e51115b304e631e3f1&vid=136f3009600b6c3abccdf5e51115b304e631e3f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR