2-Quinoxalinecarboxylic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI America
Q0081-1G
Q0081-5G
TCQ0081-1GEA
140.62
CAD
TCQ0081-1G
TCQ0081-5G
2-Quinoxalinecarboxylic acid ≥96.0% (by GC, titration analysis)
2-Quinoxalinecarboxylic acid
CAS Number: 879-65-2
MDL Number: MFCD00012334
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 209
Formula:
C₉H₆N₂O₂ Melting Pt: 209 °C |
MDL Number:
MFCD00012334 CAS Number: 879-65-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...