ɑ-Quinquethiophene

Supplier: TCI America

Q0079-100MG Q0079-500MG
TCQ0079-100MGEA 115.66 CAD
TCQ0079-100MG TCQ0079-500MG
ɑ-Quinquethiophene
alpha-Quinquethiophene

CAS Number: 5660-45-7
Molecular Formula: C20H12S5
Molecular Weight: 412.62
Form: Crystal
Color: Pale Yellow
Melting point (°C): 253

Formula: C₂₀H₁₂S₅
Melting Pt: 253 °C
CAS Number: 5660-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18f154605cfa22216a363a7fe95d33a1a8694440&vid=18f154605cfa22216a363a7fe95d33a1a8694440&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR