4-((1H-1,2,4-Triazol-1-yl)methyl)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2148-1G
A2148-5G
TCA2148-1GEA
26.86
CAD
TCA2148-1G
TCA2148-5G
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline ≥98.0% (by GC, titration analysis)
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline
CAS Number: 119192-10-8
MDL Number: MFCD00973306
Molecular Formula: C9H10N4
Molecular Weight: 174.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 128
Other Product Names / Synonyms: 1-(4-Aminobenzyl)-1,2,4-triazole
Formula:
C₉H₁₀N₄ MW: 174.21 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00973306 CAS Number: 119192-10-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...