Quincoridine ≥95.0%

Supplier: TCI America

Synonyms: (1S,2R,5R)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2R,5R)-5-Vinyl-2-quinuclidinemethanol

Q0076-1G Q0076-100MG
TCQ0076-1GEA 690.46 CAD
TCQ0076-1G TCQ0076-100MG
Quincoridine ≥95.0%
Quincoridine

CAS Number: 207129-36-0
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 267
Flash Point (°C): 122
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: 195 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (1S,2R,5R)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2R,5R)-5-Vinyl-2-quinuclidinemethanol

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Boiling Pt: 267 °C
Density: 1.05 (20 °C)
Flash Pt: 122 °C
CAS Number: 207129-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bab4247f5eaace9077433d26628943b3a942ada&vid=2bab4247f5eaace9077433d26628943b3a942ada&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR