Quinacridone ≥93.0% (by total nitrogen basis)

Supplier: TCI America

Q0057-25G Q0057-5G
TCQ0057-025GEA 104.28 CAD
TCQ0057-025G TCQ0057-5G
Quinacridone ≥93.0% (by total nitrogen basis)
Quinacridone

CAS Number: 1047-16-1
MDL Number: MFCD00059956
Molecular Formula: C20H12N2O2
Molecular Weight: 312.33
Purity/Analysis Method: >93.0% (N)
Form: Crystal
Color: Red
Melting point (°C): 390
Lambda max.: 598 nm (H2SO4)


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Quinacridone Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: 5,12-Dihydroquino[2,3-b]acridine-7,14-dione, Pigment Violet 19

Formula: C₂₀H₁₂N₂O₂
Melting Pt: 390 °C
MDL Number: MFCD00059956
CAS Number: 1047-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94aa561c505f62fa96275fa6c703ef2badcd1b43&vid=94aa561c505f62fa96275fa6c703ef2badcd1b43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR