1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone ≥90.0% (by HPLC)

Supplier: TCI America

Q0022-5G Q0022-25G
TCQ0022-005GEA 159.58 CAD
TCQ0022-005G TCQ0022-025G
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone ≥90.0% (by HPLC)
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone

CAS Number: 81-48-1
MDL Number: MFCD00019148
Molecular Formula: C21H15NO3
Molecular Weight: 329.36
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: Deep Yellow Red
Lambda max.: 583 nm (Toluene)


Other Product Names / Synonyms: 1-Hydroxy-4-p-toluidinoanthraquinone

Formula: C₂₁H₁₅NO₃
MDL Number: MFCD00019148
CAS Number: 81-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94d47dcac47307d57a694704cce9883c8355b364&vid=94d47dcac47307d57a694704cce9883c8355b364&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR